2-Propanol ≥99,7% für die Gaschromatographie

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

40983.K2 40983.M1
40983.K2EA 118 CHF
40983.K2 40983.M1
2-Propanol ≥99,7% für die Gaschromatographie
2-Propanol

Purity: 99.7+% CAS No.: 67-63-0 Molecular Formula: (CH3)2CHOH Formula Weight: 60.10 Flash Point: 12°(53°F) Physical Form: Liquid Density: 0.785 Refractive Index: 1.3770 Odour: Alcohol-like Melting Point: -90° Boiling Point: 82-83° Beilstein Reference: 635639 Merck Reference: 14,5208 UN No.: 1219 MDL No.: MFCD00011674


Suitable for environmental, pesticide, and GC analysis

Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Siedepunkt: 82…83 °C
Melting Pt: –90 °C
Dichte: 0.785
Flash-Pt: 12 °C (53 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 14,5208
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR