Kieselgel (7734) pore size 60 Å, 70-230 mesh, hochrein, Supelco®

Lieferant: Merck

403598-500G
403598-500GEA 624 CHF
403598-500G
Kieselgel (7734) pore size 60 Å, 70-230 mesh, hochrein, Supelco®
Silica gel
Formel: SiO₂
MW: 60.08 g/mol
Siedepunkt: 2230 °C
Melting Pt: > 1600 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9734803eb9ad6660974e3a900c9b4b2be516714&vid=85fd9324ea90fab0fed88ee16ec8fb766d6eca03&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR