Tetraethylorthosilikat (TEOS) ≥99,9%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Tetraethoxysilane

40251.22 40251.A3
40251.22EA 101 CHF
40251.22 40251.A3
Tetraethylorthosilikat (TEOS) ≥99,9%
Tetraethylorthosilikat (TEOS)

Purity: 99.9% CAS No.: 78-10-4 Molecular Formula: C8H20O4Si Formula Weight: 208.33 Flash Point: 46°(116°F) Physical Form: Liquid Density: 0.934 Refractive Index: 1.3820 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pleasant Melting Point: -85° Boiling Point: 168° Beilstein Reference: 1422225 Merck Reference: 14,3851 UN No.: 1292 MDL No.: MFCD00009062

Formel: Si(OC₂H₅)₄
MW: 208.33 g/mol
Siedepunkt: 168 °C
Melting Pt: –85 °C
Dichte: 0.934
Flash-Pt: 46 °C (116 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009062
CAS-Nummer: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3 III
Merck Index: 13,03882

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR