Kupfer(II)trifluormethansulfonat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Copper(II) trifluoromethanesulphonate, Copper triflate

40133.09 40133.18 40133.22
40133.09EA 214 CHF
40133.09 40133.18 40133.22
Kupfer(II)trifluormethansulfonat ≥99%
Kupfer(II)trifluormethansulfonat

Purity: 99% min CAS No.: 34946-82-2 Molecular Formula: Cu(CF3SO3)2 Formula Weight: 361.68 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to pale blue Melting Point: dec. Beilstein Reference: 4028198 UN No.: 3261 MDL No.: MFCD00077492

Formel: C₂CuF₆O₆S₂
MW: 361.69 g/mol
Melting Pt: Zersetzung
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00077492
CAS-Nummer: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8 III

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR