Natriumdihydrogenphosphat Monohydrat 99,0-102,0, crystals, ultrapure_bioreagent LC, Molecular_Biology, J.T.Baker®

Lieferant: Avantor

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

4011-01EA 202 CHF
4011-01 4011-05
Natriumdihydrogenphosphat Monohydrat 99,0-102,0, crystals, ultrapure_bioreagent LC, Molecular_Biology, J.T.Baker®
Natriumdihydrogenphosphat Monohydrat

For liquid chromatography and molecular biology applications. Lot analysis on label.

Formel: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Dichte: 2.04 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 99.0 - 102.0 %
Assay (NaH₂PO₄·H₂O) 99.0 - 102.0 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
pH of 5% Solution at 25°C 4.1 - 4.5
Insoluble Matter, Calcium and NH₄OH Precipitate ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR