Mouse IgG2b Isotype Control [clone: MPC-11]
Lieferant: Tonbo Biosciences
40-4732-U100
40-4732-U100EA 98.6 CHF
40-4732-U100
Mouse IgG2b Isotype Control [clone: MPC-11]
Antikörper
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.