Kaliumbromid für die Spektroskopie

Lieferant: Thermo Fisher Scientific
Achtung

39796.P2 39796.Q4
39796.P2EA 1920 CHF
39796.P2 39796.Q4
Kaliumbromid für die Spektroskopie
Kaliumbromid

CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: Random Cuttings, Chunks Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 730° Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Formel: KBr
Siedepunkt: 1435 °C
Melting Pt: 730 °C
Dichte: 2.75
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011358
CAS-Nummer: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR