Kupfer ≥99,999% (Metall-Basis), Pulver -100 mesh

Lieferant: Thermo Fisher Scientific
Achtung

39682.06 39682.14 39682.22
39682.06EA 168 CHF
39682.06 39682.14 39682.22
Kupfer ≥99,999% (Metall-Basis), Pulver -100 mesh
Copper

Purity: 99.999% CAS No.: 7440-50-8 Appearance: Red brown Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965

Formel: Cu
MW: 63.55 g/mol
Siedepunkt: 2595
Melting Pt: 1083
Dichte: 8.94 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010965
CAS-Nummer: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR