Praseodymtriiodid, ultra trocken ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

39385.03 39385.06
39385.03EA 258 CHF
39385.03 39385.06
Praseodymtriiodid, ultra trocken ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymtriiodid

Purity: 99.9+% CAS No.: 13813-23-5 Molecular Formula: I3Pr Formula Weight: 521.62 Physical Form: Crystalline/Needles, Ampouled under argon Sensitivity: Light Sensitive and Hygroscopic MDL No.: MFCD00049513

Formel: I₃Pr
MW: 521.62 g/mol
Melting Pt: 737 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00049513
CAS-Nummer: 13813-23-5
EINECS: 237-466-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR