Kulturkolben nach Kolle, DURAN®
Lieferant: DWK Life Sciences
215214106
391-0340EA 92.8 CHF
391-0340
Kulturkolben nach Kolle, DURAN®
Kolben Kulturkolben
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Oval neck
Typical application: preparation of cultures in nutrient media.
Order Now
TECHNISCHE DATEN
Environmentally Preferable