VWR®, Medienflaschen mit Verschluss
Lieferant: VWR Collection
391-0283EA 246 CHF
391-0283 391-0284 391-0286 391-0299
VWR®, Medienflaschen mit Verschluss
Flaschen Zellkulturflaschen
PS, PC oder PETG, mit PP-Schraubkappen, steril.
  • Sloped design improves bottle stability when placed on a work surface
  • Narrow bottle shoulder, combined with moulded finger grips, improves the ability to securely grip the bottle with gloved hands
  • Easy Grasp bottle cap facilitates uncapping, tightening and bottle stacking, as well as providing a convenient gripping surface while transporting
  • Polystyrene (PS) bottles are supplied with a white cap
  • Polycarbonate (PC) bottles are supplied with a blue cap
  • PETG bottles are supplied with a green cap
Die runden,More Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable