CO₂ incubators with hot air sterilisation, CB-S Solid.Line series
Lieferant: Binder
9640-0001 9640-0003 9640-0029
390-1608EA 9640 CHF
390-1608 390-1609 390-1673
CO₂ incubators with hot air sterilisation, CB-S Solid.Line series
Brutschränke CO₂-Brutschränke
The durable and affordable CB-S series incubators are suitable for routine cell cultivation applications in the field of cell production, particularly in academic settings. They are contamination proof due to the 180 °C hot air sterilisation cycle and reliably pH stable thanks to drift-free infrared CO₂ measurements conducted in real time.
  • Tried and tested anti-contamination concept with 180 °C hot air sterilisation
  • Humidification system with condensation protection
  • Internal data logger, measured values can be read out in open format via USB
  • Tightly fitted