Benzylmagnesiumchlorid 1.4 M THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

389551000
389551000.EA 92.9 CHF
389551000.
Benzylmagnesiumchlorid 1.4 M THF, AcroSeal®
Benzylmagnesiumchlorid
Formel: C₇H₇ClMg
MW: 150.89 g/mol
Flash-Pt: –17 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000469
CAS-Nummer: 6921-34-2
EINECS: 230-039-0
UN: 2924
ADR: 3 II

Order Now

Spezifikation Testergebnisse

Appearance Clear green-brown to brown-purple Solution (May develop some turbidity or precipitate)
Titration Grignard 19 to 21 % (As BMC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR