Zinn ≥99,5%, Granulat ACS

Lieferant: Thermo Fisher Scientific

36691.22 36691.36
36691.22EA 109 CHF
36691.22 36691.36
Zinn ≥99,5%, Granulat ACS
Zinn

Purity: 99.9% CAS No.: 7440-31-5 Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Siedepunkt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Dichte: 7.31 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00133862
CAS-Nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR