Bis(4-hydroxy-N-methylanilinium)sulfat 99,0-101,5% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt

36674.18 36674.30
36674.18EA 85.8 CHF
36674.18 36674.30
Bis(4-hydroxy-N-methylanilinium)sulfat 99,0-101,5% ACS
4-Methylaminophenolsulfat

Purity: 99.0-101.5% CAS No.: 55-55-0 Molecular Formula: C14H18N2O2.H2SO4 Formula Weight: 344.38 Physical Form: Powder Sensitivity: Air and Light Sensitive Appearance: White to pale cream Melting Point: ca. 260° dec. Beilstein Reference: 3919382 Merck Reference: 14,6017 UN No.: 2811 MDL No.: MFCD00013140

Formel: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
Siedepunkt: 227
Melting Pt: ∼. 260 °C (Zers.)
Dichte: 1.434 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013140
CAS-Nummer: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1 III
Merck Index: 14,6017

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR