Natriumcarbonat, wasserfrei 99,5-100,5% (durch Trockensubstanz) NF, ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyms: Calcined soda, Carbonic acid disodium salt

3642-26EA 0 CHF
3642-26
Natriumcarbonat, wasserfrei 99,5-100,5% (durch Trockensubstanz) NF, ACS, J.T.Baker®
Natriumcarbonat
Formel: Na₂CO₃
MW: 105.99 g/mol
Siedepunkt: 1600
Melting Pt: 854
MDL Number: MFCD00003494
CAS-Nummer: 497-19-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Meets N.F. Requirements
Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) ≤0.5 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
ACS - Assay (Na₂CO₃) (dried basis)(by acidimetry) ≥99.5 %
ACS - Calcium (Ca) ≤0.03 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤5 ppm
ACS - Loss on Heating at 285°C ≤1.0 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Silica (SiO₂) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR