Lithiumfluorid 99,85 Metals basis

Lieferant: Thermo Fisher Scientific
Gefahr

36359.18 36359.09
36359.18EA 378 CHF
36359.18 36359.09
Lithiumfluorid 99,85 Metals basis
Lithium fluoride

Purity: 99.85% CAS No.: 7789-24-4 Molecular Formula: FLi Formula Weight: 25.94 Physical Form: Lump/Powder Density: 2.635 Refractive Index: 1.3915 Appearance: White Odour: Odourless Melting Point: 845° Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090

Formel: FLi
MW: 25.94 g/mol
Siedepunkt: 1680 °C
Melting Pt: 845 °C
Dichte: 2.635
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011090
CAS-Nummer: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,5531

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR