Aluminiumfluorid, wasserfrei ≥99,5%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Aluminum fluoride

36335.14 36335.22 36335.06
36335.14EA 763 CHF
36335.14 36335.22 36335.06
Aluminiumfluorid, wasserfrei ≥99,5%
Aluminium fluoride

Purity: 99.5% CAS No.: 7784-18-1 Molecular Formula: AlF3 Formula Weight: 83.98 Physical Form: 3-12mm Sintered Lump Density: 2.882 Melting Point: 1291° subl. Merck Reference: 14,339 MDL No.: MFCD00003426


For vacuum deposition

Formel: AlF₃
MW: 83.98 g/mol
Melting Pt: 1291 °C (Subl.)
Dichte: 2.882
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003426
CAS-Nummer: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8 II
Merck Index: 14,339

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR