Lithiumsulfat Monohydrat ≥99,0% ACS

Lieferant: Thermo Fisher Scientific
Achtung

36216.36 36216.A3 36216.22
36216.36EA 357 CHF
36216.36 36216.A3 36216.22
Lithiumsulfat Monohydrat ≥99,0% ACS
Lithiumsulfat Monohydrat

Purity: 99.0% min CAS No.: 10102-25-7 Molecular Formula: Li2SO4.H2O Formula Weight: 127.96 (109.94anhy) Physical Form: Crystalline Density: 2.06 Melting Point: 130° -H2O Solubility: Soluble in water Merck Reference: 14,5541 MDL No.: MFCD00149766

Formel: Li₂SO₄·H₂O
Melting Pt: 130 °C (-H₂O)
Dichte: 2.06
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149766
CAS-Nummer: 10102-25-7
EINECS: 233-820-4
Merck Index: 14,5541

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR