Human processed serum (HPS), TSH depleted

Lieferant: SEQENS
36000D-100ML 36000D-500ML 36000D-1L
36000D-100MLEA 557 CHF
36000D-100ML 36000D-500ML 36000D-1L
Human processed serum (HPS), TSH depleted
Seren
SEQENS IVD manufactures TSH Depleted Human Processed Serum.

  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • TSH: <0,1 UI/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years

For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

Bitte beachten Only for research or manufacture of in vitro diagnostic reagents. Not forMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR