Eisen(II)chlorid, ultra trocken ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Iron dichloride, Ferrous chloride

Unter Argon in Ampullen verpackt

35701.03 35701.09 35701.18
35701.03EA 143 CHF
35701.03 35701.09 35701.18
Eisen(II)chlorid, ultra trocken ≥99,99% (Metall-Basis)
Eisen(II)chlorid

Purity: 99.99% CAS No.: 7758-94-3 Molecular Formula: Cl2Fe Formula Weight: 126.75 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.160 pH: 2.5 Melting Point: 674° Boiling Point: 1023° Merck Reference: 14,4043 UN No.: 3260 MDL No.: MFCD00011004

Formel: Cl₂Fe
MW: 126.75 g/mol
Siedepunkt: 1023 °C
Melting Pt: 674
Dichte: 3.160
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011004
CAS-Nummer: 7758-94-3
EINECS: 231-843-4
UN: 3260
ADR: 8 III
Merck Index: 14,4043

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR