Kaliumcarbonat 98, powder -325 mesh, reagent

Lieferant: Honeywell Chemicals
Achtung

Synonyms: Potash

347825-1KG 347825-2.5KG 347825-25KG
347825-1KGEA 191.55 CHF
347825-1KG 347825-2.5KG HONC347825-25KG
Kaliumcarbonat 98, powder -325 mesh, reagent
Kaliumcarbonat

  • ≥95% for general laboratory use.

Formel: K₂CO₃
MW: 138.21 g/mol
Siedepunkt: Zersetzung
Melting Pt: 891 °C
Dichte: 2.430 g/cm³ (19 °C)
Lager Temperatur: Room Temperature
MDL Number: MFCD00011382
CAS-Nummer: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Form powder
Assay 98.0 - 102.0 %
Particle size distribution 50 % Max. 44 µm
Particle Size −325 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR