Natriumacetat Trihydrat 99,0-101,0% (Trockenmasse), Kristalle USP, Multikompendial, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyms: Acetic acid sodium salt trihydrate

Management of Change (MOC) category = HR

3461-01EA 114 CHF
3461-01 3461-05 3461-07 3461-08 3461-19
Natriumacetat Trihydrat 99,0-101,0% (Trockenmasse), Kristalle USP, Multikompendial, J.T.Baker®
Natriumacetat Trihydrat

Optimize biopharmaceutical fermentation and cell culture productivity and improve the efficiency of your pharmaceutical production and biopharmaceutical processes with our proven-effective vitamins and pharmaceutical process minerals.


  • Food GMP manufactured product

Formel: H₃CCOONa·3H₂O
Siedepunkt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Dichte: 1.45 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00071557
CAS-Nummer: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Meets U.S.P Requirements 99.0 - 101.0%
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements ≤0.035%
Food GMP Manufactured Product ≤2.5
Meets J.P. Chemical Specifications Passes Test
Meets E.P. Chemical Specifications ≤0.05%
Meets B.P. Chemical Specifications 38.0 - 41.0%
Preserve in tight containers, store at room temperature. 7.5 - 9.2
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical ≤0.005%
Bulk Pharmaceutical Chemical 99.0 - 101.0%
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤ 0.035 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
USP - Identification Passes Test
USP - Insoluble Matter ≤ 0.05 %
USP - Loss on Drying 38.0 - 41.0 %
USP - pH of 5% Solution at 25°C 7.5 - 9.2
USP - Potassium (K) Passes Test
USP - Sulfate (SO₄) ≤ 0.005 %
EP/BP - Assay (CH₃COONa) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH (1 in 20) 7.5 - 9.0
EP/BP - Reducing Substances Passes Test
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 200 ppm
EP/BP - Calcium and Magnesium ≤ 50 ppm
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying 39.0 - 40.5 %
JP - Assay (CH₃COONa) 99.5 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Chloride (Cl) ≤ 0.011 %
JP - Sulfate (SO₄) ≤ 0.017 %
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Calcium and Magnesium Passes Test
JP - Arsenic (As) ≤ 2 ppm
JP - Substances Reducing Permanganate Passes Test
JP - Loss on Drying 39.0 - 40.5 %
FCC - Alkalinity ≤ 0.05 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Loss on Drying 38.0 - 41.0 %
FCC - Potassium Compounds Passes Test
No Class 1, 2, 3 or other solvent is likely to be present in the final
product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR