Human processed serum (HPS), hormone depleted

Lieferant: SEQENS
34000D-100ML 34000D-500ML 34000D-1L
34000D-100MLEA 354 CHF
34000D-100ML 34000D-500ML 34000D-1L
Human processed serum (HPS), hormone depleted
Seren
SEQENS IVD manufactures Hormones Depleted Human Processed Serum.

  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Cortisol: <0,11 µg/dl
  • Total T3: <0,6 nmol/L
  • Total T4: <13 nmol/L
  • Free T3: <1,5 pmol/L
  • Free T4: <5,2 pmol/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years

For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceabilityMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR