Supelco® PESTANAL® Organic Reference Standard, Bromocyclen

Lieferant: Merck

33398-100MG
33398-100MGEA 62.1 CHF
33398-100MG
Supelco® PESTANAL® Organic Reference Standard, Bromocyclen
Standards Organische Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fa16873504f7d961a3d71f64c63b9fc93aca166&vid=9fa16873504f7d961a3d71f64c63b9fc93aca166&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

M-Clarity Qualitätsstufe MQ100
Synonym(s) 5- Bromomethyl-1,2,3,4,7,7-hexachloro-2-norbornene
Environmentally Preferable
CAS Nr. 1715-40-8
MDL No. MFCD00151378
Formel C₈H₅BrCl₆
Molecular weight 393,75
Flammpunkt Not applicable
Shipping conditions STD
Lagerbedingungen 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR