Dimethylgelb Indikator, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: Dimethyl yellow, Sudan Yellow, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene

33129-25G
33129-25GEA 127.31 CHF
33129-25G
Dimethylgelb Indikator, Fluka™
Dimethylgelb
Formel: C₁₄H₁₅N₃
MW: 225.29 g/mol
Siedepunkt: 371
Melting Pt: 111 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008308
CAS-Nummer: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1 III
Merck Index: 12,03279

Order Now

Spezifikation Testergebnisse

Identity (IR) complying
Change complying
Extinction maximum at 403 - 413 nm
Absorbtion (1%/1cm) 1050 - 1350


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR