3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyms: 3-Hexyl-2-[3-(3-hexyl-1,3-benzoxazol-2(3H)-ylidene)-1-propen-1-yl]-1,3-benzoxazol-3-ium iodide, DiOC6(3) iodide, DiOC6(3), 3,3'-Dihexyloxacarbocyanine iodide, 3-Hexyl-2-[3-(3-hexyl-2(3H)benzoxazolylidene)-1-propenyl]benzoxazolium iodide

M-Clarity™ Qualitätsstufe = MQ100

318426-1G 318426-250MG
318426-1GEA 593 CHF
318426-1G 318426-250MG
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

3,3′-dihexyloxacarbocyanine iodide (DiOC6) is a supravital lipophilic fluorochrome. This cationic dye is employed to stain mitochondria and endoplasmic reticulum in animal and plant cells. In addition, it acts as a very rapid, consistent and highly selective fluorochrome for endophytic ascomycetes in plant roots and liverworts. DiOC6 also facilitates the detection of different mitochondrial changes happening during early apoptosis.


  • 3,3′-Dihexyloxacarbocyanine iodide has been used
  • to selectively stain endophytic ascomycetes in plant roots and liverworts
  • to visualize thrombus formation and platelet aggregate
  • to visualize forchlorfenuron (FCF)-induced changes in mitochondrial morphology

Formel: C₂₉H₃₇IN₂O₂
MW: 572.531 g/mol
Melting Pt: 219...221 °C (Lit.)
Lager Temperatur: Room Temperature
MDL Number: MFCD00011954
CAS-Nummer: 53213-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c42b6b5dcf8cee0d894d078ece1453c15591058&vid=fbaed696ad598fe13c40b56852a8e6d8fd1093f5&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Colour) Red
Appearance (Form) Powder or Chunks
Infrared spectrum Conforms to Structure
Wavelength 483 - 488 nm
Extinction Coefficient, c = 0.01 g/L; Methanol ≥120000
Carbon 59.3 - 62.4%
Nitrogen 4.6 - 5.1%
Purity (HPLC) ≥97.5%
Solubility (Turbidity) Clear to Slightly Hazy
Solubility (Colour), c = 25 mg/ml, CHCl3 Orange

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR