Methylorange 0,04% aqueous solution, TECHNICAL Indicator_pH

Lieferant: VWR Chemicals

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

31720.292EA 60.6 CHF
31720.292 31720.230
Methylorange 0,04% aqueous solution, TECHNICAL Indicator_pH
Methylorange

  • pH-Bereich des Farbwechsels: 3,1-4,4

Formel: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Siedepunkt: 90
Flash-Pt: 38
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007502
CAS-Nummer: 547-58-0
EINECS: 208-925-3
Merck Index: 13,06127

Order Now

Spezifikation Testergebnisse

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR