Centrifuge tubes, SuperClear®
Lieferant: LABCON
3191-870-008-9 3192-870-008-9 3131-165-008-9 3131-325-008-9 3131-340-008-9 3131-345-008-9 3132-340-008-9 3132-345-008-9 3141-330-018-9 3141-335-018-9 3141-870-008-9 3142-330-018-9 3142-335-018-9 3142-870-008-9 3181-165-008-9 3181-325-008-9 3181-340-008-9 3181-345-008-9 3182-340-008-9 3182-345-008-9 3191-330-018-9 3191-335-018-9 3192-330-018-9 3192-335-018-9 3191-335-028-9
LBCN3191-870-008-9EA 282 CHF
LBCN3191-870-008-9 3192-870-008-9 3131-165-008-9 3131-325-008-9 3131-340-008-9 3131-345-008-9 3132-340-008-9 3132-345-008-9 3141-330-018-9 3141-335-018-9 3141-870-008-9 3142-330-018-9 3142-335-018-9 3142-870-008-9 3181-165-008-9 3181-325-008-9 3181-340-008-9 3181-345-008-9 3182-340-008-9 3182-345-008-9 3191-330-018-9 3191-335-018-9 3192-330-018-9 3192-335-018-9 3191-335-028-9
Centrifuge tubes, SuperClear®
Röhrchen Zentrifugenröhrchen
SuperClear® high performance centrifuge tubes are lot-tested at a much higher g-force than rated to ensure that they can be spun safely. The quality control team tests the seals with chloroform. 
  • ATP-, RNase/DNase- and endotoxin-free (non-pyrogenic)
  • Patented anti-smear writing surface
  • 3 colour printed graduations and reverse reading scale
  • Convenient freeze line
  • Ultra-clear Polypropylene USP Class VI resin, free of DiHEMDA, oleamide, and BPA
  • Freezable to –80 °C, and autoclavable
SuperClear® tubes are made from an ultra-clear, medical grade resinMore Product Information