qPCR master mixes, Fast EvaGreen®
Lieferant: Biotium
31003-1 31003-2 31003 31003-T
31003-1EA 669 CHF
31003-1 31003-2 31003. 31003-T
qPCR master mixes, Fast EvaGreen®
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.
  • 2X qPCR master mix for fast cycling protocols
  • EvaGreen® dye-based
  • Contains Cheetah™ Taq hotstart DNA polymerase
Fast Evagreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.
Fast Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.