4-Amino-3,6-disulpho-1,8-naphthalic anhydride dipotassium salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

307122-100MG
307122-100MGEA 874 CHF
307122-100MG
4-Amino-3,6-disulpho-1,8-naphthalic anhydride dipotassium salt, Sigma-Aldrich®
4-Amino-3,6-disulpho-1,8-naphthalic anhydride dipotassium salt
Formel: C₁₂H₅K₂NO₉S₂
MW: 449.50 g/mol
MDL Number: MFCD00012400
CAS-Nummer: 79539-35-8

Order Now

Spezifikation Testergebnisse

Appearance (Colour) Faint Orange to Very Dark Orange
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon 30.3 - 33.8%
Wavelength 348 - 352 nm
Extinction Coefficient ≥20000
Extinction Coefficient, c = 0.01 g/L, 0.1N NaOH ≥8000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR