Trifluoressigsäure 99,0, CHROMASOLV™ HPLC, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: TFA

302031-100ML 302031-1L 302031-10X1ML
302031-100MLEA 347.48 CHF
302031-100ML 302031-1L 302031-10X1ML
Trifluoressigsäure 99,0, CHROMASOLV™ HPLC, Fluka™
Trifluoressigsäure

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Beilstein Number: 742035

Formel: C₂HF₃O₂
MW: 114.02 g/mol
Siedepunkt: 72 °C (1.013 hPa)
Melting Pt: –15 °C
Dichte: 1.48 g/cm³ (20 °C)
Flash-Pt: 100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004169
CAS-Nummer: 76-05-1
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay (alkalimetric) 99.0 - 101.0 %
Assay (GC) Min. 99.0 %
Identity (IR) complying
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Absorbance at 320 nm Max. 0.025
Absorbance at 300 nm Max. 0.03
Absorbance at 290 nm Max. 0.04
Absorbance 280 nm Max. 0.05
Absorbance at 270 nm Max. 0.10
Absorbance at 260 nm Max. 0.9
APHA Max. 20
Vapor Density 3.9 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR