Anti-IgE Goat Polyclonal Antibody (Biotin)

Lieferant: Tonbo Biosciences
30-8027-M001
30-8027-M001EA 504 CHF
30-8027-M001
Anti-IgE Goat Polyclonal Antibody (Biotin)
Antikörper
Goat anti-Human IgE (epsilon chain) - Affinity Pure, Biotin Conjugate, min x w/bovine, mouse or rabbit serum proteins

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: ε chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR