Diethylpyrocarbonat, Millipore®

Lieferant: Merck Millipore Calbiochem
Achtung

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

298711-100 298711-25
298711-100EA 1070 CHF
298711-100 298711-25
Diethylpyrocarbonat, Millipore®
Diethylpyrocarbonat
Formel: O(COOC₂H₅)₂
Siedepunkt: 93 °C (1013 hPa)
Dichte: 1.123 g/cm³ (20 °C)
Flash-Pt: 69 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00009106
CAS-Nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR