Polyvinylalkohol, M.W. 115,000 88, hydrolysed

Lieferant: VWR Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

297914DEA 133 CHF
297914D
Polyvinylalkohol, M.W. 115,000 88, hydrolysed
Polyvinylalkohol
Formel: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS-Nummer: 9002-89-5

Order Now

Spezifikation Testergebnisse

Appearance White granular powder
pH (4 %) 4.5 - 6.5
Viscosity (20°C; 4 %; water) 46 - 54 mPa.s
Degree of hydrolysis 86.5 - 89 %
Ignition residue Max. 0.5 %
Loss on drying (60°C) Max. 5 %
Organic volatile impurities Max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR