Polypropylenglycol, M.W. 2,000

Lieferant: VWR Chemicals

Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG

297776TEA 385 CHF
297776T
Polypropylenglycol, M.W. 2,000
Polypropylenglycol
Formel: H(OCH₂CH₂CH₂)nOH
Dichte: 1.005 g/cm³ (20 °C)
Flash-Pt: 112.8 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00084446
CAS-Nummer: 25322-69-4
EINECS: 500-039-8

Order Now

Spezifikation Testergebnisse

Identification by IR Passes test
Colouration Max. 20 APHA
Hydroxyl value 53 - 58
Viscosity (25°C) 300 - 340 mPa.s
Water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR