D(+)-Xylose, GPR RECTAPUR®

Lieferant: VWR Chemicals

29013.237EA 195 CHF
29013.237
D(+)-Xylose, GPR RECTAPUR®
D(+)-Xylose
Formel: C₅H₁₀O₅
Siedepunkt: 331 °C (1013 hPa)
Melting Pt: 154 °C
Dichte: 1.525 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00151475
CAS-Nummer: 58-86-6
EINECS: 200-400-7
Merck Index: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Melting point 145 - 150 °C
Specific optical rotation (10 %; water) 18 - 20 °
Ignition residue (SO4) Max. 0.2 %
Loss on drying (100°C) Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR