Eosin B 1 g/l aqueous solution, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91

MQ500

2853-100ML
2853-100MLEA 111 CHF
2853-100ML
Eosin B 1 g/l aqueous solution, Sigma-Aldrich®
Eosin B
Formel: C₂₀H₆Br₂N₂Na₂O₉
MW: 624.06 g/mol
MDL Number: MFCD00005041
CAS-Nummer: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR