Natriumdihydrogenphosphat 98, GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

28013.264EA 125 CHF
28013.264 28013.366
Natriumdihydrogenphosphat 98, GPR RECTAPUR®
Natriumdihydrogenphosphat
Formel: NaH₂PO₄
MW: 119.98 g/mol
Melting Pt: ∼650 °C
Dichte: 0.9996 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003527
CAS-Nummer: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Spezifikation Testergebnisse

Assay Min. 98 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR