Oxalsäure Dihydrat 99,5-102,5, crystals, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyms: Ethanedioic acid dihydrate

2752-20EA 0 CHF
2752-20
Oxalsäure Dihydrat 99,5-102,5, crystals, AR® ACS, Macron Fine Chemicals™
Oxalsäure Dihydrat
Formel: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 101
MDL Number: MFCD00149102
CAS-Nummer: 6153-56-6
UN: 3261
ADR: 8 III
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.5 - 102.5 %
ACS - Assay (H₂C₂O₄2H₂O) 99.5 - 102.5 %
ACS - Calcium (Ca) ≤ 0.001 %
ACS - Chloride (Cl) ≤ 0.002 %
ACS - Heavy Metals (as Pb) ≤ 0.0005 %
ACS - Insoluble Matter ≤ 0.005 %
ACS - Iron (Fe) ≤ 0.0002 %
ACS - Nitrogen Compounds (as N) ≤ 0.001 %
ACS - Residue after Ignition ≤ 0.01 %
Solubility Passes Test
ACS - Substances Darkened by Hot H₂SO₄ Passes Test
ACS - Sulfate (SO₄) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR