Kernechtrot, TECHNICAL

Lieferant: VWR Chemicals

Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL

27416.101EA 272 CHF
27416.101
Kernechtrot, TECHNICAL
Kernechtrot
Formel: C₁₄H₈NNaO₇S
MW: 357.28 g/mol
Melting Pt: 300
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00078493
CAS-Nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512

Order Now

Spezifikation Testergebnisse

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR