Kaliumsulfat 99,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Lieferant: VWR Chemicals

26997.293EA 50.3 CHF
26997.293 26997.460
Kaliumsulfat 99,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Potassium sulphate
Formel: K₂SO₄
MW: 174.26 g/mol
Siedepunkt: 1698
Melting Pt: 1069 °C
Dichte: 2.66 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011388
CAS-Nummer: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Spezifikation Testergebnisse

Assay Min. 99.0 %
pH (25°C; 5 %) 5.5 - 7.5
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 100 ppm
Total N (Nitrogen) Max. 5 ppm
Cl (Chloride) Max. 5 ppm
As (Arsenic) Max. 2 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 20 ppm
Na (Sodium) Max. 0.02 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR