Kaliumcarbonat 99,0, AnalaR NORMAPUR® Reag. Ph. Eur. zA

Lieferant: VWR Chemicals
Achtung

Synonyms: Potash

26726.297EA 141 CHF
26726.297 26726.322 26726.260 26726.460
Kaliumcarbonat 99,0, AnalaR NORMAPUR® Reag. Ph. Eur. zA
Kaliumcarbonat
Formel: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Dichte: 2.42 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011382
CAS-Nummer: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.0 %
Ba (Barium) Passes test
CN (Cyanide) Passes test
Sulphur compounds (as S) Passes test
Free potassium hydroxide Max. 0.05 %
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Substances precipitated by NH4OH Max. 100 ppm
Cl (Chloride) Max. 30 ppm
NO3 (Nitrate) Max. 30 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 + SO3 (as SO4) Max. 20 ppm
As (Arsenic) Max. 2 ppm
Ca + Mg (as Ca) Max. 100 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 0.2 %
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR