Kaliumhydroxid 38% aqueous solution, AnalaR NORMAPUR® zA

Lieferant: VWR Chemicals
Gefahr

Synonyms: Potash caustic, Caustic potash

26632.293EA 70.1 CHF
26632.293
Kaliumhydroxid 38% aqueous solution, AnalaR NORMAPUR® zA
Kaliumhydroxid
Formel: KOH
MW: 56.11 g/mol
Siedepunkt: 1320
Melting Pt: 360
Dichte: 1.38 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003553
CAS-Nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Spezifikation Testergebnisse

Assay 38.0 - 40.3 %
Density (20/4) 1.375 - 1.400
Heavy metals (as Pb) Max. 2 ppm
Potassium carbonate Max. 1.0 %
Silica Max. 10 ppm
Total N (Nitrogen) Max. 1 ppm
Cl (Chloride) Max. 50 ppm
PO4 (Phosphate) Max. 1 ppm
SO4 (Sulphate) Max. 2 ppm
Al (Aluminium) Max. 1 ppm
Ba (Barium) Max. 1 ppm
Ca (Calcium) Max. 5 ppm
Cd (Cadmium) Max. 0.1 ppm
Co (Cobalt) Max. 0.1 ppm
Cr (Chromium) Max. 2 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 1 ppm
Hg (Mercury) Max. 0.01 ppm
Mg (Magnesium) Max. 1 ppm
Mn (Manganese) Max. 0.1 ppm
Na (Sodium) Max. 0.25 %
Ni (Nickel) Max. 2 ppm
Pb (Lead) Max. 0.5 ppm
Zn (Zinc) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR