2-Desoxy-D-glucose, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

25972-1
25972-1EA 157 CHF
25972-1
2-Desoxy-D-glucose, Millipore®
2-Deoxy-D-glucose
Formel: C₆H₁₂O₅
MDL Number: MFCD00151328
CAS-Nummer: 154-17-6
EINECS: 205-823-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR