Mangan(II)chlorid Tetrahydrat 98,0-101,0 EDTA titration, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

2540-07EA 0 CHF
2540-07 2540-01 2540-R 2540-05
Mangan(II)chlorid Tetrahydrat 98,0-101,0 EDTA titration, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Mangan(II)chlorid Tetrahydrat
Formel: MnCl₂·4H₂O
MW: 197.9 g/mol
Siedepunkt: 1190 °C (1013 hPa)
Melting Pt: 58…59 °C
Dichte: 2.01 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149792
CAS-Nummer: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9 III
Merck Index: 13,05751

Order Now

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 98.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
ACS - Assay (MnCl₂·4H₂O) (by EDTA titrn) 98.0 - 101.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 3.5 - 6.0
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
ACS - Zinc (Zn) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR