7-Amino-4-(trifluormethyl)cumarin ≥99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyms: 7-Amino-4-(trifluoromethyl)coumarin, 7-Amino-4-trifluoromethylcoumarin, 7-Amino-4-(trifluoromethyl)-2H-chromen-2-one, Coumarin 151

M-Clarity™ quality level = MQ100

248924-1G 248924-250MG
248924-1GEA 187 CHF
248924-1G 248924-250MG
7-Amino-4-(trifluormethyl)cumarin ≥99%, Sigma-Aldrich®
7-Amino-4-(trifluormethyl)cumarin

7-Amino-4-(trifluoromethyl)coumarin (AFC) is a novel fluorescent marker used for the detection of proteinases. Suitable as laser dye.

Formel: C₁₀H₆F₃NO₂
Melting Pt: 221...222 °C (lit.)
Lager Temperatur: Room Temperature
MDL Number: MFCD00006858
CAS-Nummer: 53518-15-3
EINECS: 258-599-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5f44c556789195f428f69406f1f321d056732b3&vid=1db6c3c283098f8289e3bf94a3de48150ef85a58&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Color) Yellow
Appearance (Form) Powder or Crystals or Chunk(s) or Powder with Chunk(s) or Crystals with Chunk(s)
Infrared spectrum Conforms to Structure
Carbon 51.9 - 52.9 %
Nitrogen 5.9 - 6.3 %
Purity (GC) ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR