Poly(chlortrifluorethylen) 650, VOLTALEF®

Lieferant: VWR Chemicals

24630.293EA 0 CHF
24630.293
Poly(chlortrifluorethylen) 650, VOLTALEF®
Poly(chlortrifluorethylen) 650
Formel: (C₂ClF₃)n
MW: 650 g/mol
CAS-Nummer: 9002-83-9

Order Now

Spezifikation Testergebnisse

Identification Passes test

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR