Florisil® 60-100 mesh Chromatography

Lieferant: VWR Chemicals

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

24278.182EA 82 CHF
24278.182 24278.295 24278.230 24278.440
Florisil® 60-100 mesh Chromatography
Magnesiumsilikat
Formel: MgO₃Si
MW: 100.38 g/mol
MDL Number: MFCD00078249
CAS-Nummer: 1343-88-0
EINECS: 215-681-1

Order Now

Spezifikation Testergebnisse

Identification Passes Test

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR