Cäsiumchlorid 99,5, AnalaR NORMAPUR® zA

Lieferant: VWR Chemicals

Synonyms: Cesium Chloride

22960.131EA 106 CHF
22960.131 22960.180
Cäsiumchlorid 99,5, AnalaR NORMAPUR® zA
Cesium chloride
Formel: CsCl
MW: 168.36 g/mol
Siedepunkt: 1303
Melting Pt: 646
Dichte: 3.7808 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010955
CAS-Nummer: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Spezifikation Testergebnisse

Assay Min. 99.5 %
Total N (Nitrogen) Max. 10 ppm
SO4 (Sulphate) Max. 20 ppm
Al (Aluminium) Max. 5 ppm
Cu (Copper) Max. 3 ppm
Fe (Iron) Max. 3 ppm
K (Potassium) Max. 20 ppm
Li (Lithium) Max. 2 ppm
Mg (Magnesium) Max. 5 ppm
Na (Sodium) Max. 20 ppm
Pb (Lead) Max. 1 ppm
Rb (Rubidium) Max. 200 ppm
Zn (Zinc) Max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR